Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207541.1 | NP_997424.1 | 4 | 439 | ||
GenBank mRNA | AK135853.1 | - | 4 | 440 | ||
BC117831.1 | - | 4 | 389 | 40092935 |