Search
Search

Cdc14a| Gene Symbol: Cdc14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080818.2 | NP_001074287.1 | 16 | 3440 | ||
| NM_001173553.1 | NP_001167024.1 | 14 | 3293 | |||
| GenBank mRNA | AK036556.1 | - | 13 | 2681 | ||
| AK142294.1 | - | 16 | 3443 | |||
| BC072644.1 | - | 14 | 3247 | 30356962 | ||