Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013378.2 | NP_001013396.2 | 9 | 1768 | ||
NM_001115149.1 | NP_001108621.1 | 9 | 1768 | |||
NM_001115150.1 | NP_001108622.1 | 8 | 1599 | |||
NM_001115151.1 | NP_001108623.1 | 8 | 1599 | |||
NM_001115153.1 | NP_001108625.1 | 10 | 1945 | |||
NM_001286824.1 | NP_001273753.1 | 9 | 1726 | |||
NM_001286825.1 | NP_001273754.1 | 9 | 1710 | |||
GenBank mRNA | AK050969.1 | - | 9 | 1752 | ||
AK088787.1 | - | 8 | 1465 | |||
AK145409.1 | - | 10 | 1843 | |||
AK146820.1 | - | 9 | 1593 | |||
AK161654.1 | - | 8 | 1446 | |||
AK168702.1 | - | 9 | 1593 | |||
AK168981.1 | - | 9 | 1620 | |||
BC028849.1 | - | 9 | 1662 | |||
BC032291.1 | - | 6 | 3738 | |||
BC086664.1 | - | 9 | 1610 |