Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175022.2 | NP_778187.2 | 14 | 6827 | ||
GenBank mRNA | AK032894.1 | - | 11 | 3315 | ||
BC058674.1 | - | 11 | 5466 | |||
BC075628.1 | - | 15 | 5737 |