Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033320.2 | NP_001028492.1 | 11 | 1563 | ||
GenBank mRNA | AK031012.1 | - | 11 | 1562 | ||
BC115830.1 | - | 3 | 183 | |||
HF678090.1 | - | 32 | 3194 |