Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbeal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183276.2 | NP_899099.2 | 26, 27 | 4015 | ||
GenBank mRNA | AK129158.1 | - | 2, 3 | 168 | ||
BC157956.1 | - | 26, 27 | 3996 | 9007284 | ||
BC172118.1 | - | 25, 26 | 3915 | 9054404 |