Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017316.2 | NP_059012.2 | 6 | 713 | ||
XM_006235067.2 | XP_006235129.1 | 6 | 1510 | |||
XM_006235068.2 | XP_006235130.1 | 7 | 1513 | |||
XM_006235069.2 | XP_006235131.1 | 4 | 2955 | |||
GenBank mRNA | AF080453.1 | - | 6 | 713 |