Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FASN SIRNA Assays ›
Gene Symbol:
Fasn
Gene Name:
fatty acid synthase
Gene Aliases:
-
Chromosome Location:
Chr.10: 109987735 - 110005901 on Build Rnor_6.0
UniGene ID:
Rn.9486
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50273
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Fasn
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017332.1
NP_059028.1
4
393
GenBank mRNA
M76767.1
-
4
393
X13415.1
-
2
191
X62888.1
-
4
398
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
acetyl-CoA metabolic process
fatty acid biosynthetic process
lipid biosynthetic process
oxidation-reduction process
cellular response to interleukin-4
fatty acid synthase activity
[acyl-carrier-protein] S-acetyltransferase activity
[acyl-carrier-protein] S-malonyltransferase activity
3-oxoacyl-[acyl-carrier-protein] synthase activity
3-oxoacyl-[acyl-carrier-protein] reductase (NADPH) activity
3-hydroxypalmitoyl-[acyl-carrier-protein] dehydratase activity
enoyl-[acyl-carrier-protein] reductase (NADPH, B-specific) activity
oleoyl-[acyl-carrier-protein] hydrolase activity
drug binding
zinc ion binding
myristoyl-[acyl-carrier-protein] hydrolase activity
palmitoyl-[acyl-carrier-protein] hydrolase activity
identical protein binding
protein homodimerization activity
poly(A) RNA binding
enoyl-[acyl-carrier-protein] reductase (NADPH, A-specific) activity
3-hydroxyoctanoyl-[acyl-carrier-protein] dehydratase activity
NADPH binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0