Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101488.1 | NP_001094958.1 | 7 | 2601 | ||
GenBank mRNA | AK090234.1 | - | 1 | 597 |