Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033421.3 | NP_001028593.1 | 4 | 2140 | ||
GenBank mRNA | AK136651.1 | - | 4 | 2141 |