Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033474.2 | NP_001028646.2 | 1 | 3182 | ||
GenBank mRNA | AK003616.1 | - | 1 | 1614 | ||
AK014824.1 | - | 1 | 3079 | |||
AK037008.1 | - | 36 | ||||
AK051537.1 | - | 1064 | ||||
AK078766.1 | - | 1 | 950 |