Search
Search

Zbtb42| Gene Symbol: Zbtb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100460.1 | NP_001093930.1 | 2 | 2152 | ||
| GenBank mRNA | AK079245.1 | - | 1 | 149 | ||
| BC023876.1 | - | 1 | 1080 | |||
| BC026524.1 | - | 1 | 412 | |||
| BC046387.1 | - | 3 | 1859 | |||
| BC049242.1 | - | 1 | 444 | |||
| BC051084.1 | - | 1 | 2178 | |||
| BC055474.1 | - | 1 | 199 | |||
| BC069952.1 | - | 1 | 186 | |||