Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277204.1 | NP_001264133.1 | 3 | 718 | ||
NM_001277205.1 | NP_001264134.1 | 4 | 665 | |||
NM_001277206.1 | NP_001264135.1 | 3 | 623 | |||
NM_001277207.1 | NP_001264136.1 | 3 | 565 | |||
NM_001277208.1 | NP_001264137.1 | 2 | 632 | |||
NM_176796.2 | NP_789766.1 | 4 | 698 | |||
NM_176797.2 | NP_789767.1 | 3 | 538 | |||
NM_176798.2 | NP_789768.1 | 4 | 815 | |||
XM_005274022.3 | XP_005274079.1 | 2 | 2806 | |||
XM_006718571.3 | XP_006718634.1 | 3 | 2891 | |||
XM_011545076.2 | XP_011543378.1 | 3 | 2021 | |||
XM_011545077.2 | XP_011543379.1 | 4 | 2140 | |||
XM_011545079.2 | XP_011543381.1 | 2 | 454 | |||
GenBank mRNA | AF007891.1 | - | 3 | 504 | ||
AF007892.1 | - | 4 | 815 | |||
AF007893.1 | - | 3 | 1339 | |||
AF218005.1 | - | 3 | 575 | |||
AF498920.1 | - | 1 | 273 | |||
AL520218.3 | - | 4 | 686 | |||
AL525099.3 | - | 4 | 660 | |||
AX775903.1 | - | 4 | 815 | |||
BC000571.2 | - | 2 | 513 | 3162734 | ||
BC009391.2 | - | 2 | 572 | 4128941 | ||
BT006771.1 | - | 1 | 273 | |||
BX337204.2 | - | 3 | 565 | |||
BX381745.2 | - | 4 | 686 | |||
BX397649.2 | - | 3 | 612 | |||
U52464.1 | - | 3 | 476 | |||
X97058.1 | - | 3 | 549 |
Set Membership: |
G-protein coupled receptor |