Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163317.1 | NP_001156789.1 | 3 | 708 | ||
GenBank mRNA | BC147370.1 | - | 3 | 707 | 9056434 | |
BC147371.1 | - | 3 | 707 | 9056432 | ||
BC147726.1 | - | 3 | 708 | 9056675 | ||
BC147731.1 | - | 3 | 708 | 9056680 |