Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otogl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177567.1 | NP_001171038.1 | 3 | 116 | ||
GenBank mRNA | GU140005.1 | - | 3 | 117 |