Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040696.1 | NP_001035786.1 | 4 | 2053 | ||
NM_001162414.1 | NP_001155886.1 | 3 | 2025 | |||
GenBank mRNA | BC141354.1 | - | 3 | 1954 | 9055966 | |
DQ117591.1 | - | 4 | 2053 | |||
DQ117592.1 | - | 3 | 1926 | |||
DQ117593.1 | - | 3 | 1926 | |||
DQ153214.1 | - | 4 | 2053 | |||
EF158039.1 | - | 3 | 2025 |