Search
Search

Nsmce4a| Gene Symbol: Nsmce4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162855.1 | NP_001156327.1 | 1, 2 | 395 | ||
| GenBank mRNA | AK010349.1 | - | 1, 2 | 396 | ||
| AK010795.1 | - | 1, 2 | 361 | |||
| BE951712.1 | - | 1, 2 | 272 | |||