Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027410.1 | NP_081686.1 | 10 | 1577 | ||
GenBank mRNA | AB093295.1 | - | 9 | 1746 | ||
AK032858.1 | - | 10 | 1551 | |||
AK034020.1 | - | 9 | 2155 | |||
AK169888.1 | - | 10 | 1524 | |||
AK170108.1 | - | 10 | 1524 | |||
AK182741.1 | - | 1 | 128 | |||
AK192321.1 | - | 1 | 128 | |||
AK196900.1 | - | 1 | 128 | |||
AK199155.1 | - | 1 | 40 | |||
AK218769.1 | - | 2 | 337 | |||
AK218802.1 | - | 1 | 128 | |||
BC046968.1 | - | 10 | 1577 | 6306206 | ||
BC052185.1 | - | 4 | 2154 |