Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146012.2 | NP_001139484.2 | 2 | 137 | ||
NM_001301651.1 | NP_001288580.1 | 2 | 89 | |||
NM_001301652.1 | NP_001288581.1 | 1 | 370 | |||
GenBank mRNA | AK006177.1 | - | 2 | 89 | ||
AK032854.1 | - | 2 | 107 | |||
AK156843.1 | - | 2 | 107 | |||
BC052536.1 | - | 1 | 286 | |||
BC059860.1 | - | 1 | 370 | |||
CJ181933.1 | - | 2 | 134 |