Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081050.2 | NP_001074519.2 | 23 | 6131 | ||
GenBank mRNA | AK007379.1 | - | 1 | 1307 |