Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028657.3 | NP_082933.2 | 2, 3 | 533 | ||
GenBank mRNA | AK149678.1 | - | 2, 3 | 539 | ||
AK154386.1 | - | 2, 3 | 535 | |||
AK154902.1 | - | 2, 3 | 613 | |||
AK170185.1 | - | 2, 3 | 553 | |||
AK171084.1 | - | 2, 3 | 535 | |||
BC118620.1 | - | 2, 3 | 533 | 40107417 | ||
BC119790.1 | - | 2, 3 | 533 | 40107416 |