Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167883.1 | NP_001161355.1 | 4 | 1682 | ||
GenBank mRNA | AK129484.1 | - | 4 | 1667 | ||
AK147106.1 | - | 2 | 1822 | |||
AK162481.1 | - | 2 | 1530 | |||
AK220375.1 | - | 2 | 1819 | |||
BC060298.1 | - | 2 | 614 | |||
BC145902.1 | - | 2 | 1501 | 40131205 |