Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002016.2 | NP_001002016.2 | 4 | 994 | ||
GenBank mRNA | BC062018.1 | - | 3 | 549 | 5621637 | |
DN934590.1 | - | 4 | 734 | |||
X66870.1 | - | 4 | 953 | |||
X76297.1 | - | 4 | 716 | |||
X99257.1 | - | 4 | 453 |