Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021869.2 | NP_068641.2 | 8, 9 | 702 | ||
XM_006227691.2 | XP_006227753.1 | 9, 10 | 2024 | |||
XM_008758626.1 | XP_008756848.1 | 9, 10 | 2018 | |||
GenBank mRNA | AF031430.1 | - | 7, 8 | 606 | ||
BC085737.1 | - | 8, 9 | 702 | 7193349 |
Gene Symbol: LOC100910446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758670.1 | XP_008756892.1 | 6, 7 | 588 |