Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022506.3 | NP_071951.1 | 3 | 198 | ||
GenBank mRNA | BC062228.1 | - | 351 | 6921741 | ||
FQ211300.1 | - | 3 | 216 | |||
FQ216848.1 | - | 3 | 197 | |||
FQ221256.1 | - | 3 | 197 | |||
FQ221939.1 | - | 3 | 196 | |||
FQ222938.1 | - | 3 | 201 | |||
FQ222963.1 | - | 3 | 197 | |||
FQ223725.1 | - | 3 | 196 | |||
FQ226734.1 | - | 196 | ||||
FQ228354.1 | - | 3 | 197 | |||
FQ228461.1 | - | 3 | 197 | |||
FQ228715.1 | - | 3 | 194 | |||
X04809.1 | - | 3 | 198 |