Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022926.1 | NP_075215.1 | 2 | 572 | ||
XM_006253083.1 | XP_006253145.1 | 2 | 597 | |||
GenBank mRNA | AF076167.1 | - | 2 | 572 |