Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKAA2 SIRNA Assays ›
Gene Symbol:
Prkaa2
Gene Name:
protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Aliases:
Ampk, Ampka2
Chromosome Location:
Chr.5: 124568845 - 124642569 on Build Rnor_6.0
UniGene ID:
Rn.64583
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
52562
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Prkaa2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_023991.1
NP_076481.1
2
130
GenBank mRNA
U12149.1
-
2
315
Z29486.1
-
2
130
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_006252, EF056019, DA710012, BC069823, BC069680, and others.
This siRNA also targets the following transcripts in Mouse: NM_178143, BC138565, BY718361, AK044030, AK134573, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
fatty acid biosynthetic process
cholesterol biosynthetic process
autophagy
lipid biosynthetic process
positive regulation of autophagy
response to activity
Wnt signaling pathway
regulation of lipid metabolic process
response to caffeine
cellular response to nutrient levels
negative regulation of TOR signaling
histone-serine phosphorylation
cellular response to glucose starvation
glucose homeostasis
regulation of circadian rhythm
negative regulation of apoptotic process
positive regulation of glycolytic process
rhythmic process
protein heterooligomerization
fatty acid homeostasis
cellular response to prostaglandin E stimulus
cellular response to organonitrogen compound
regulation of energy homeostasis
chromatin binding
protein kinase activity
protein serine/threonine kinase activity
AMP-activated protein kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
protein binding, bridging
histone serine kinase activity
metal ion binding
[hydroxymethylglutaryl-CoA reductase (NADPH)] kinase activity
[acetyl-CoA carboxylase] kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0