Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023976.2 | NP_076466.2 | 7 | 4013 | ||
XM_006228586.2 | XP_006228648.1 | 7 | 4106 | |||
XM_008759196.1 | XP_008757418.1 | 6 | 4207 | |||
GenBank mRNA | Z29649.2 | - | 7 | 4013 |