Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GCH1 SIRNA Assays ›
Gene Symbol:
Gch1
Gene Name:
GTP cyclohydrolase 1
Gene Aliases:
Gch
Chromosome Location:
Chr.15: 23935011 - 23968971 on Build Rnor_6.0
UniGene ID:
Rn.28195
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
52785
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gch1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024356.1
NP_077332.1
1
291
GenBank mRNA
FQ220953.1
-
1
278
M58364.1
-
1
291
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001024070, NM_001024071, NM_000161, NM_001024024, S44049, and others.
Panther Classification:
Molecular Function -
hydrolase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
tetrahydrobiopterin biosynthetic process
metabolic process
regulation of blood pressure
regulation of lung blood pressure
response to lipopolysaccharide
response to interferon-gamma
response to tumor necrosis factor
7,8-dihydroneopterin 3'-triphosphate biosynthetic process
vasodilation
dopamine biosynthetic process
pteridine-containing compound biosynthetic process
negative regulation of blood pressure
tetrahydrofolate biosynthetic process
response to pain
neuromuscular process controlling posture
positive regulation of nitric-oxide synthase activity
dihydrobiopterin metabolic process
protein homooligomerization
protein heterooligomerization
GTP cyclohydrolase I activity
calcium ion binding
protein binding
GTP binding
zinc ion binding
GTP-dependent protein binding
protein homodimerization activity
coenzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0