Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031111.1 | NP_112373.1 | 4 | 173 | ||
XM_006235823.1 | XP_006235885.1 | 4 | 262 | |||
GenBank mRNA | BC058464.1 | - | 4 | 178 | 6918340 | |
FQ210654.1 | - | 4 | 176 | |||
FQ210736.1 | - | 4 | 176 | |||
FQ210823.1 | - | 4 | 174 | |||
FQ210868.1 | - | 4 | 175 | |||
FQ212305.1 | - | 4 | 175 | |||
FQ212673.1 | - | 4 | 168 | |||
FQ217300.1 | - | 4 | 174 | |||
FQ221031.1 | - | 4 | 176 | |||
FQ221222.1 | - | 4 | 176 | |||
FQ221352.1 | - | 4 | 174 | |||
FQ221376.1 | - | 4 | 174 | |||
FQ221504.1 | - | 4 | 174 | |||
FQ221719.1 | - | 4 | 175 | |||
FQ221773.1 | - | 4 | 174 | |||
FQ221882.1 | - | 4 | 174 | |||
FQ221895.1 | - | 4 | 174 | |||
FQ222195.1 | - | 4 | 174 | |||
FQ222197.1 | - | 4 | 174 | |||
FQ222346.1 | - | 4 | 175 | |||
FQ222395.1 | - | 4 | 174 | |||
FQ222640.1 | - | 4 | 177 | |||
FQ222661.1 | - | 4 | 174 | |||
FQ222692.1 | - | 4 | 176 | |||
FQ222717.1 | - | 4 | 174 | |||
FQ222861.1 | - | 4 | 174 | |||
FQ222985.1 | - | 4 | 174 | |||
FQ223083.1 | - | 4 | 174 | |||
FQ223085.1 | - | 4 | 174 | |||
FQ223126.1 | - | 4 | 174 | |||
FQ223140.1 | - | 4 | 173 | |||
FQ223281.1 | - | 4 | 174 | |||
FQ223324.1 | - | 4 | 174 | |||
FQ223458.1 | - | 4 | 175 | |||
FQ223473.1 | - | 4 | 204 | |||
FQ224512.1 | - | 4 | 175 | |||
FQ224728.1 | - | 4 | 176 | |||
FQ228462.1 | - | 4 | 174 | |||
FQ228565.1 | - | 4 | 175 | |||
FQ228580.1 | - | 4 | 175 | |||
FQ228593.1 | - | 4 | 174 | |||
FQ228604.1 | - | 4 | 179 | |||
FQ229072.1 | - | 4 | 169 | |||
FQ229301.1 | - | 4 | 178 | |||
FQ229813.1 | - | 4 | 174 | |||
X79059.1 | - | 4 | 173 |