Search Thermo Fisher Scientific
Gene Symbol: Rab3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031091.2 | NP_112353.1 | 5 | 621 | ||
XM_006238531.2 | XP_006238593.1 | 5 | 694 | |||
GenBank mRNA | BC060562.1 | - | 5 | 566 | 6918181 | |
U68184.1 | - | 4 | 327 | |||
Y14019.1 | - | 4 | 509 |