Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR1I3 SIRNA Assays ›
          Gene Symbol:
          NR1I3
          Gene Name:
          nuclear receptor subfamily 1 group I member 3
          Gene Aliases:
          CAR, CAR1, MB67
          Chromosome Location:
          Chr.1: 161229666 - 161238623 on Build GRCh38
          UniGene ID:
          Hs.349642
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5358

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR1I3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077469.2 NP_001070937.1 2, 3 318
          NM_001077470.2 NP_001070938.1 1, 2 178
          NM_001077471.2 NP_001070939.1 2, 3 318
          NM_001077472.2 NP_001070940.1 1, 2 178
          NM_001077473.2 NP_001070941.1 1, 2 178
          NM_001077474.2 NP_001070942.1 2, 3 318
          NM_001077475.2 NP_001070943.1 1, 2 178
          NM_001077476.2 NP_001070944.1 1, 2 178
          NM_001077477.2 NP_001070945.1 1, 2 178
          NM_001077478.2 NP_001070946.1 2, 3 318
          NM_001077479.2 NP_001070947.1 1, 2 178
          NM_001077480.2 NP_001070948.1 2, 3 318
          NM_001077481.2 NP_001070949.1 2, 3 318
          NM_001077482.2 NP_001070950.1 2, 3 318
          NM_005122.4 NP_005113.1 2, 3 318
          XM_005245693.4 XP_005245750.1 1, 2 591
          XM_005245694.4 XP_005245751.1 1, 2 590
          XM_005245697.4 XP_005245754.1 2, 3 201
          XM_011510237.2 XP_011508539.1 1, 2 588
          GenBank mRNA AK303757.1 - 1, 2 171
          AK303796.1 - 2, 3 352
          AK310773.1 - 1, 2 59
          AK316208.1 - 2, 3 310
          AY572806.1 - 2, 3 209
          AY572808.1 - 1, 2 69
          AY572809.1 - 1, 2 69
          AY572810.1 - 2, 3 209
          AY572811.1 - 1, 2 69
          AY572812.1 - 1, 2 69
          AY572813.1 - 2, 3 209
          AY572814.1 - 1, 2 69
          AY572815.1 - 1, 2 69
          AY572816.1 - 1, 2 69
          AY572817.1 - 1, 2 69
          AY572818.1 - 1, 2 69
          AY572819.1 - 2, 3 209
          AY572820.1 - 1, 2 69
          AY572822.1 - 2, 3 209
          AY572823.1 - 2, 3 209
          AY572824.1 - 1, 2 69
          AY572825.1 - 1, 2 69
          AY572826.1 - 2, 3 209
          AY572827.1 - 1, 2 69
          BC069626.1 - 2, 3 271 7262456
          BC121120.1 - 2, 3 219 40120664
          BC121121.1 - 2, 3 219 40120665
          CB124867.1 - 2, 3 318
          DQ022681.1 - 1, 2 115
          HQ692838.1 - 1, 2 115
          HQ692839.1 - 1, 2 115
          HQ692840.1 - 1, 2 115
          HQ692841.1 - 1, 2 115
          HQ709178.1 - 1, 2 115

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          transcription initiation from RNA polymerase II promoter
          signal transduction
          androgen receptor signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          androgen receptor activity
          thyroid hormone receptor activity
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0