Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031521.1 | NP_113709.1 | 3 | 366 | ||
XM_006242980.2 | XP_006243042.1 | 3 | 517 | |||
XM_006242986.2 | XP_006243048.1 | 3 | 517 | |||
XM_006242988.2 | XP_006243050.1 | 3 | 517 | |||
XM_006242994.2 | XP_006243056.1 | 3 | 517 | |||
XM_008766192.1 | XP_008764414.1 | 3 | 517 | |||
XM_008766193.1 | XP_008764415.1 | 3 | 517 | |||
GenBank mRNA | BC101924.1 | - | 3 | 367 | 7936825 | |
X06564.1 | - | 3 | 366 |