Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POR SIRNA Assays ›
Gene Symbol:
Por
Gene Name:
P450 (cytochrome) oxidoreductase
Gene Aliases:
CYPOR
Chromosome Location:
Chr.12: 23998411 - 24017063 on Build Rnor_6.0
UniGene ID:
Rn.11359
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
53838
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Por
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031576.1
NP_113764.1
986
GenBank mRNA
M10068.1
-
8, 9
950
M12516.1
-
986
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008898, BC003240, BC031463, D17571.
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of growth plate cartilage chondrocyte proliferation
response to nutrient
carnitine metabolic process
flavonoid metabolic process
internal peptidyl-lysine acetylation
fatty acid oxidation
positive regulation of chondrocyte differentiation
positive regulation of monooxygenase activity
response to drug
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
nitrate catabolic process
positive regulation of cholesterol biosynthetic process
positive regulation of smoothened signaling pathway
nitric oxide catabolic process
oxidation-reduction process
negative regulation of lipase activity
demethylation
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to peptide hormone stimulus
positive regulation of steroid hormone biosynthetic process
regulation of cholesterol metabolic process
cellular organofluorine metabolic process
NADPH-hemoprotein reductase activity
cytochrome-b5 reductase activity, acting on NAD(P)H
iron ion binding
nitric oxide dioxygenase activity
electron carrier activity
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
hydrolase activity
enzyme binding
iron-cytochrome-c reductase activity
flavin adenine dinucleotide binding
NADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0