Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031595.1 | NP_113783.1 | 8 | 1079 | ||
GenBank mRNA | BC062019.1 | - | 8 | 989 | 5620971 | |
D83522.1 | - | 8 | 953 | |||
FQ225567.1 | - | 7 | 738 | |||
FQ225947.1 | - | 7 | 738 | |||
FQ226273.1 | - | 7 | 738 | |||
FQ232987.1 | - | 7 | 739 | |||
FQ233836.1 | - | 7 | 738 | |||
U77918.1 | - | 8 | 1079 |