Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031732.2 | NP_113920.2 | 3 | 311 | ||
GenBank mRNA | BC088125.1 | - | 3 | 311 | 7324981 | |
FQ219411.1 | - | 3 | 341 | |||
L22339.1 | - | 2 | 221 |