Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053456.1 | NP_445908.1 | 1, 2 | 717 | ||
GenBank mRNA | D45920.1 | - | 1, 2 | 717 |