Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053653.1 | NP_446105.1 | 6 | 1078 | ||
GenBank mRNA | AY032729.1 | - | 6 | 1078 |