Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS15A SIRNA Assays ›
          Gene Symbol:
          Rps15a
          Gene Name:
          ribosomal protein S15a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 187759855 - 187766734 on Build Rnor_6.0
          UniGene ID:
          Rn.2890
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 56051

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps15a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053982.1 NP_446434.1 1 100
          XM_006230097.1 XP_006230159.1 2 147
          XM_006230098.1 XP_006230160.1 2 190
          GenBank mRNA BC058452.1 - 2 125 6921126
          FQ210209.1 - 2 122
          FQ211215.1 - 2 125
          FQ212042.1 - 2 121
          FQ212257.1 - 2 119
          FQ214481.1 - 2 121
          FQ220927.1 - 2 117
          FQ221112.1 - 2 129
          FQ221144.1 - 2 152
          FQ221174.1 - 2 119
          FQ221230.1 - 2 194
          FQ221308.1 - 2 123
          FQ221419.1 - 2 121
          FQ221518.1 - 2 121
          FQ221677.1 - 2 119
          FQ221720.1 - 2 190
          FQ221825.1 - 2 121
          FQ221840.1 - 2 121
          FQ221877.1 - 2 121
          FQ222252.1 - 2 132
          FQ222272.1 - 2 121
          FQ222274.1 - 2 121
          FQ222304.1 - 2 121
          FQ222376.1 - 2 121
          FQ222415.1 - 2 119
          FQ222418.1 - 2 123
          FQ222718.1 - 2 121
          FQ222721.1 - 2 119
          FQ222768.1 - 2 119
          FQ222892.1 - 2 122
          FQ222921.1 - 2 124
          FQ222922.1 - 2 190
          FQ223006.1 - 2 121
          FQ223095.1 - 2 121
          FQ223230.1 - 2 120
          FQ223341.1 - 2 121
          FQ223389.1 - 2 121
          FQ223433.1 - 2 121
          FQ223787.1 - 2 121
          FQ223822.1 - 2 120
          FQ223930.1 - 243
          FQ224201.1 - 2 121
          FQ224347.1 - 2 121
          FQ224383.1 - 2 121
          FQ228419.1 - 2 120
          FQ228429.1 - 2 121
          FQ228446.1 - 2 121
          FQ228475.1 - 2 122
          FQ228508.1 - 2 121
          FQ228609.1 - 2 121
          FQ228617.1 - 2 120
          FQ229004.1 - 2 122
          FQ229042.1 - 2 119
          FQ229063.1 - 2 120
          X77953.1 - 1 100

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          positive regulation of cell proliferation
          response to virus
          positive regulation of cell cycle
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0