Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014913.3 | NP_055728.1 | 2, 3 | 559 | ||
XM_005266656.4 | XP_005266713.1 | 2, 3 | 413 | |||
GenBank mRNA | AB020670.1 | - | 2, 3 | 289 | ||
AK292566.1 | - | 2, 3 | 328 | |||
BC071589.1 | - | 2, 3 | 559 | 30333923 |