siRNA Details
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277249.1 | NP_001264178.1 | 4 | 346 | ||
NM_001277250.1 | NP_001264179.1 | 4 | 346 | |||
NM_001277251.1 | NP_001264180.1 | 5 | 405 | |||
NM_001277252.1 | NP_001264181.1 | 4 | 286 | |||
NM_001277253.1 | NP_001264182.1 | 4 | 398 | |||
NM_057143.2 | NP_476484.1 | 4 | 528 | |||
GenBank mRNA | AF157511.1 | - | 4 | 528 | ||
AF157512.1 | - | 4 | 282 | |||
AJ007291.1 | - | 5 | 453 | |||
BC091128.1 | - | 4 | 319 | 7376914 | ||
FM055257.1 | - | 4 | 316 | |||
FM061417.1 | - | 4 | 267 | |||
FM069664.1 | - | 4 | 337 | |||
FQ220895.1 | - | 4 | 398 | |||
FQ223564.1 | - | 4 | 396 | |||
FQ224480.1 | - | 5 | 389 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -






