Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS15A SIRNA Assays ›
          Gene Symbol:
          Rps15a
          Gene Name:
          ribosomal protein S15a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 187759855 - 187766734 on Build Rnor_6.0
          UniGene ID:
          Rn.2890
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 56234

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps15a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053982.1 NP_446434.1 2, 3 220
          XM_006230097.1 XP_006230159.1 3, 4 267
          XM_006230098.1 XP_006230160.1 3, 4 310
          GenBank mRNA BC058452.1 - 3, 4 245 6921126
          FQ211215.1 - 3, 4 245
          FQ212042.1 - 3, 4 241
          FQ212257.1 - 3, 4 239
          FQ214481.1 - 3, 4 241
          FQ220418.1 - 1, 2 65
          FQ220927.1 - 3, 4 237
          FQ220997.1 - 3, 4 241
          FQ221041.1 - 3, 4 241
          FQ221112.1 - 3, 4 249
          FQ221144.1 - 3, 4 272
          FQ221174.1 - 3, 4 239
          FQ221230.1 - 3, 4 314
          FQ221308.1 - 3, 4 243
          FQ221419.1 - 3, 4 241
          FQ221518.1 - 3, 4 241
          FQ221677.1 - 3, 4 239
          FQ221720.1 - 3, 4 310
          FQ221825.1 - 3, 4 241
          FQ221840.1 - 3, 4 241
          FQ221877.1 - 3, 4 241
          FQ222252.1 - 3, 4 252
          FQ222272.1 - 3, 4 241
          FQ222274.1 - 3, 4 241
          FQ222304.1 - 3, 4 241
          FQ222376.1 - 3, 4 241
          FQ222415.1 - 3, 4 239
          FQ222418.1 - 3, 4 243
          FQ222718.1 - 3, 4 241
          FQ222721.1 - 3, 4 239
          FQ222768.1 - 3, 4 239
          FQ222892.1 - 3, 4 242
          FQ222922.1 - 3, 4 310
          FQ223006.1 - 3, 4 241
          FQ223095.1 - 3, 4 241
          FQ223230.1 - 3, 4 240
          FQ223341.1 - 3, 4 241
          FQ223389.1 - 3, 4 241
          FQ223433.1 - 3, 4 241
          FQ223787.1 - 3, 4 241
          FQ223822.1 - 3, 4 240
          FQ223930.1 - 363
          FQ224201.1 - 3, 4 241
          FQ224347.1 - 3, 4 241
          FQ224383.1 - 3, 4 241
          FQ228419.1 - 3, 4 240
          FQ228429.1 - 3, 4 241
          FQ228446.1 - 3, 4 241
          FQ228475.1 - 3, 4 242
          FQ228508.1 - 3, 4 241
          FQ228609.1 - 3, 4 241
          FQ228617.1 - 3, 4 240
          FQ229004.1 - 3, 4 242
          FQ229042.1 - 3, 4 239
          FQ229063.1 - 3, 4 240
          X77953.1 - 2, 3 220

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          positive regulation of cell proliferation
          response to virus
          positive regulation of cell cycle
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9hqbx:80/100.66.78.73:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0