Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOA SIRNA Assays ›
          Gene Symbol:
          Rhoa
          Gene Name:
          ras homolog family member A
          Gene Aliases:
          Arha, Arha2
          Chromosome Location:
          Chr.8: 117082440 - 117116244 on Build Rnor_6.0
          UniGene ID:
          Rn.107401
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 56274

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rhoa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_057132.3 NP_476473.1 3 365
          XM_006243699.1 XP_006243761.1 5 633
          XM_006243700.1 XP_006243762.1 5 608
          XM_006243701.1 XP_006243763.1 5 728
          GenBank mRNA AY026068.1 - 3 365
          AY026069.1 - 3 365
          BC061732.1 - 5 542 5598621
          D84477.1 - 3 335

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001664, L25080, AK307220, AF498970, AK130066, and others.

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          response to hypoxia
          regulation of transcription from RNA polymerase II promoter
          cytoskeleton organization
          cell adhesion
          cell-matrix adhesion
          small GTPase mediated signal transduction
          skeletal muscle tissue development
          regulation of actin polymerization or depolymerization
          metabolic process
          response to mechanical stimulus
          response to glucose
          regulation of neuron projection development
          substantia nigra development
          cerebral cortex cell migration
          forebrain radial glial cell differentiation
          actin cytoskeleton organization
          cell differentiation
          positive regulation of cell growth
          regulation of cell migration
          positive regulation of cell migration
          androgen receptor signaling pathway
          positive regulation of actin filament polymerization
          stress-activated protein kinase signaling cascade
          positive regulation of cytokinesis
          negative regulation of intracellular steroid hormone receptor signaling pathway
          regulation of osteoblast proliferation
          cleavage furrow formation
          apolipoprotein A-I-mediated signaling pathway
          response to drug
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          stress fiber assembly
          response to amino acid
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          apical junction assembly
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          ossification involved in bone maturation
          wound healing, spreading of cells
          response to ethanol
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          neuron projection morphogenesis
          regulation of dendrite development
          negative chemotaxis
          response to glucocorticoid
          positive regulation of stress fiber assembly
          regulation of calcium ion transport
          negative regulation of cell death
          trabecula morphogenesis
          cellular response to progesterone stimulus
          positive regulation of podosome assembly
          mitotic spindle assembly
          skeletal muscle satellite cell migration
          regulation of neural precursor cell proliferation
          GTPase activity
          protein binding
          GTP binding
          myosin binding
          GDP binding
          protein domain specific binding
          Rho GDP-dissociation inhibitor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0