Search
Search

PTPN23| Gene Symbol: PTPN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304482.1 | NP_001291411.1 | 3 | 319 | ||
| NM_015466.3 | NP_056281.1 | 4 | 447 | |||
| XM_005265031.2 | XP_005265088.2 | 4 | 428 | |||
| GenBank mRNA | AB025194.1 | - | 4 | 395 | ||
| AB040904.2 | - | 4 | 447 | |||
| AF290614.1 | - | 4 | 393 | |||
| AK055157.1 | - | 4 | 418 | |||
| AK289502.1 | - | 4 | 396 | |||
| AK295877.1 | - | 4 | 367 | |||
| AK295888.1 | - | 3 | 340 | |||
| AK299906.1 | - | 3 | 290 | |||
| BC089042.1 | - | 4 | 365 | 6579163 | ||
| DA093189.1 | - | 4 | 428 | |||