Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101672.1 | NP_001095142.1 | 3, 4 | 1726 | ||
NM_001102454.1 | NP_001095924.1 | 3, 4 | 1716 | |||
NM_016423.2 | NP_057507.2 | 3, 4 | 1860 | |||
XM_005267739.1 | XP_005267796.1 | 3, 4 | 1671 | |||
XM_006720163.2 | XP_006720226.1 | 3, 4 | 1607 | |||
XM_006720164.3 | XP_006720227.1 | 3, 4 | 1609 | |||
XM_011536825.1 | XP_011535127.1 | 3, 4 | 1681 | |||
XM_017021354.1 | XP_016876843.1 | 2, 3 | 1676 | |||
XM_017021355.1 | XP_016876844.1 | 3, 4 | 1572 | |||
GenBank mRNA | AB015427.1 | - | 3, 4 | 1850 | ||
AK056261.1 | - | 1, 2 | 1067 | |||
AK225811.1 | - | 3, 4 | 1623 | |||
AK290700.1 | - | 3, 4 | 1726 | |||
BC000694.2 | - | 2, 3 | 1471 | 3347777 | ||
BC036105.1 | - | 3, 4 | 1552 | 5274736 | ||
BT006956.1 | - | 1, 2 | 1433 |