Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130421.2 | NP_569105.1 | 5, 6 | 447 | ||
GenBank mRNA | AB072980.1 | - | 5, 6 | 287 | ||
BC100066.1 | - | 5, 6 | 447 | 7387128 | ||
FQ232596.1 | - | 5, 6 | 461 |