siRNA Details
Gene Symbol: Taf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133615.1 | NP_598299.1 | 4, 5 | 413 | ||
XM_006256993.2 | XP_006257055.1 | 5, 6 | 880 | |||
XM_006256994.2 | XP_006257056.1 | 5, 6 | 879 | |||
GenBank mRNA | BC090332.1 | - | 4, 5 | 468 | 7193160 | |
U40188.1 | - | 4, 5 | 413 |

More Information
Other Species Targeted

Panther Classification:
