Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134326.2 | NP_599153.2 | 3 | 227 | ||
GenBank mRNA | BC085359.1 | - | 3 | 227 | 7303856 | |
FQ209504.1 | - | 3 | 249 | |||
FQ209784.1 | - | 3 | 246 | |||
FQ210189.1 | - | 3 | 246 | |||
FQ210310.1 | - | 3 | 245 | |||
FQ210436.1 | - | 3 | 246 | |||
FQ211062.1 | - | 3 | 250 | |||
FQ211231.1 | - | 3 | 246 | |||
FQ211307.1 | - | 3 | 246 | |||
FQ211332.1 | - | 3 | 246 | |||
FQ218069.1 | - | 3 | 249 | |||
FQ218125.1 | - | 3 | 246 | |||
FQ219734.1 | - | 3 | 242 | |||
V01222.1 | - | 3 | 212 |