Search
Search

Slc6a13| Gene Symbol: Slc6a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133623.1 | NP_598307.1 | 3 | 447 | ||
| XM_006237218.2 | XP_006237280.1 | 3 | 431 | |||
| XM_006237219.2 | XP_006237281.1 | 4 | 547 | |||
| XM_006237220.2 | XP_006237282.1 | 3 | 431 | |||
| XM_006237221.2 | XP_006237283.1 | 4 | 547 | |||
| XM_006237222.2 | XP_006237284.1 | 2 | 285 | |||
| XM_006237223.2 | XP_006237285.1 | 2 | 430 | |||
| GenBank mRNA | M95762.1 | - | 3 | 447 | ||