Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138527.1 | NP_612536.1 | 4, 5 | 559 | ||
GenBank mRNA | AB022883.1 | - | 4, 5 | 559 | ||
AF226607.1 | - | 4, 5 | 526 | |||
FQ220484.1 | - | 4, 5 | 564 | |||
FQ230176.1 | - | 4, 5 | 564 |